Product Details

SNP ID
rs72780665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:44967617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAGACTTTTATATTATTTATTC[C/T]TGCTGTATGTGCACAGCTTTGGAAG
Phenotype
MIM: 611309 MIM: 610559
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf10 PubMed Links

Gene Details

Gene
C10orf10
Gene Name
chromosome 10 open reading frame 10
There are no transcripts associated with this gene.

Gene
RASSF4
Gene Name
Ras association domain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032023.3 Intron NP_114412.2
XM_005271833.4 Intron XP_005271890.1
XM_005271835.4 Intron XP_005271892.1
XM_006718013.3 Intron XP_006718076.1
XM_006718014.3 Intron XP_006718077.1
XM_011540255.2 Intron XP_011538557.1
XM_017016754.1 Intron XP_016872243.1
XM_017016755.1 Intron XP_016872244.1
XM_017016756.1 Intron XP_016872245.1
XM_017016757.1 Intron XP_016872246.1
XM_017016758.1 Intron XP_016872247.1
Gene
TMEM72-AS1
Gene Name
TMEM72 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details