Product Details

SNP ID
rs72776700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:100808616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAATAGTAAAAAATAAATTAAAA[G/T]ATTAAAATTGTGAATATATATTTTC
Phenotype
MIM: 602547
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR548P PubMed Links

Gene Details

Gene
MIR548P
Gene Name
microRNA 548p
There are no transcripts associated with this gene.

Gene
ST8SIA4
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005668.5 4641 UTR 3 NP_005659.1
NM_175052.2 4641 Intron NP_778222.1
XM_005272078.2 4641 Intron XP_005272135.1
XM_011543630.1 4641 UTR 3 XP_011541932.1

View Full Product Details