Product Details

SNP ID
rs73348510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:2624048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTTTATAAGTCTTCATGGGCTG[C/T]TGGGGGGATAGATTCCACATCCCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01600 PubMed Links

Gene Details

Gene
LINC01600
Gene Name
long intergenic non-protein coding RNA 1600
There are no transcripts associated with this gene.

View Full Product Details