Product Details

SNP ID
rs74041956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101274868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCACATAGAGCAGCTTTGGTTTT[G/T]GTACACTATTAAAGAAAGCAAAATG
Phenotype
MIM: 603521 MIM: 607918
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SNRPA1 PubMed Links

Gene Details

Gene
SNRPA1
Gene Name
small nuclear ribonucleoprotein polypeptide A'
There are no transcripts associated with this gene.

Gene
VIMP
Gene Name
VCP interacting membrane selenoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018445.5 Intron NP_060915.2
NM_203472.2 Intron NP_982298.2

View Full Product Details