Product Details

SNP ID
rs74123852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:8046813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCCCATTGGGTGTCGGGTCGCA[C/T]AGGGGGTCTGATTATGGAGTCTTTC
Phenotype
MIM: 131320
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GATA3 PubMed Links

Gene Details

Gene
GATA3
Gene Name
GATA binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002295.1 Intron NP_001002295.1
NM_002051.2 Intron NP_002042.1
XM_005252442.2 Intron XP_005252499.1
XM_005252443.4 Intron XP_005252500.1
Gene
GATA3-AS1
Gene Name
GATA3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details