Product Details

SNP ID
rs73493927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7729945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGAGTGAGGAAGCCCTAGAAAG[A/G]AGGGGACATCTGAGCCTGCAGAGTC
Phenotype
MIM: 616256
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLEC4G PubMed Links

Gene Details

Gene
CLEC4G
Gene Name
C-type lectin domain family 4 member G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244856.1 Intron NP_001231785.1
NM_198492.3 Intron NP_940894.1

View Full Product Details