Product Details

SNP ID
rs74412712
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:3665039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGTTAAACAGGAGATGCAAACT[C/T]CAAGCTAGTTAATGAGCTGTGATCG
Phenotype
MIM: 601990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TP73 PubMed Links

Gene Details

Gene
TP73
Gene Name
tumor protein p73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126240.2 Intron NP_001119712.1
NM_001126241.2 Intron NP_001119713.1
NM_001126242.2 Intron NP_001119714.1
NM_001204184.1 Intron NP_001191113.1
NM_001204185.1 Intron NP_001191114.1
NM_001204186.1 Intron NP_001191115.1
NM_001204187.1 Intron NP_001191116.1
NM_001204188.1 Intron NP_001191117.1
NM_001204189.1 Intron NP_001191118.1
NM_001204190.1 Intron NP_001191119.1
NM_001204191.1 Intron NP_001191120.1
NM_001204192.1 Intron NP_001191121.1
NM_005427.3 Intron NP_005418.1

View Full Product Details