Product Details

SNP ID
rs78289163
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:150648763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGTCTCACCTATGAGAAAAAAG[A/G]AATAGGACATAATGAACTGAAAGAG
Phenotype
MIM: 612208
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLPH3L PubMed Links

Gene Details

Gene
GOLPH3L
Gene Name
golgi phosphoprotein 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018178.5 Intron NP_060648.2
XM_006711428.1 Intron XP_006711491.1

View Full Product Details