Product Details

SNP ID
rs75692754
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:202879101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGATACGTATATAAAAGTTTCAC[C/T]AGAAATAAAATAAATGCAAATTAAA
Phenotype
MIM: 603417
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC148709 PubMed Links

Gene Details

Gene
LOC148709
Gene Name
actin pseudogene
There are no transcripts associated with this gene.

Gene
RABIF
Gene Name
RAB interacting factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002871.4 2408 UTR 3 NP_002862.2

View Full Product Details