Product Details

SNP ID
rs75449619
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:203641535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCCCCTAAATAAGAGGGCATGC[A/G]AGTTGTCCCAGAATCTGTGGATTCA
Phenotype
MIM: 108732
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP2B4 PubMed Links

Gene Details

Gene
ATP2B4
Gene Name
ATPase plasma membrane Ca2+ transporting 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001396.2 Intron NP_001001396.1
NM_001684.4 Intron NP_001675.3

View Full Product Details