Product Details

SNP ID
rs78635036
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:155904534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAACAAGGGTCATTATGTATTGA[C/T]GCAATCTAGCCCAACTACACACACA
Phenotype
MIM: 609591
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0907 PubMed Links

Gene Details

Gene
KIAA0907
Gene Name
KIAA0907
There are no transcripts associated with this gene.

Gene
RIT1
Gene Name
Ras like without CAAX 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256820.1 Intron NP_001243749.1
NM_001256821.1 Intron NP_001243750.1
NM_006912.5 Intron NP_008843.1

View Full Product Details