Product Details

SNP ID
rs78351078
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:156410913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTTGTATCGCTTATCTCTTGGC[A/G]TCCTCCAGGAAGCCTTCCTGAGCTC
Phenotype
MIM: 611186
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf61 PubMed Links

Gene Details

Gene
C1orf61
Gene Name
chromosome 1 open reading frame 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320453.1 Intron NP_001307382.1
NM_001320454.1 Intron NP_001307383.1
NM_001320455.1 Intron NP_001307384.1
NM_006365.2 Intron NP_006356.1
XM_011509067.1 Intron XP_011507369.1
XM_011509068.1 Intron XP_011507370.1
XM_011509070.1 Intron XP_011507372.1
XM_011509071.2 Intron XP_011507373.1
XM_011509072.1 Intron XP_011507374.1
XM_011509073.2 Intron XP_011507375.1
XM_011509074.1 Intron XP_011507376.1
XM_017000061.1 Intron XP_016855550.1
XM_017000062.1 Intron XP_016855551.1
XM_017000063.1 Intron XP_016855552.1
XM_017000064.1 Intron XP_016855553.1
XM_017000065.1 Intron XP_016855554.1
XM_017000066.1 Intron XP_016855555.1
Gene
MIR9-1
Gene Name
microRNA 9-1
There are no transcripts associated with this gene.

View Full Product Details