Product Details

SNP ID
rs77899304
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:23630085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAAGTGTGAATTGCTATTACCC[C/T]AGTGTGAGGCAAGTCCTTTGACTCC
Phenotype
MIM: 607305
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MDS2 PubMed Links

Gene Details

Gene
MDS2
Gene Name
myelodysplastic syndrome 2 translocation associated
There are no transcripts associated with this gene.

View Full Product Details