Product Details

SNP ID
rs77010008
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:25863297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGCCAATGCAGGAAAGCCAGGC[A/G]AGAAAGGCAGCCATGGGCAGAAAAA
Phenotype
MIM: 607779
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AUNIP PubMed Links

Gene Details

Gene
AUNIP
Gene Name
aurora kinase A and ninein interacting protein
There are no transcripts associated with this gene.

Gene
PAQR7
Gene Name
progestin and adipoQ receptor family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178422.5 1032 Silent Mutation CTC,CTT L181L NP_848509.1
XM_005245745.3 1032 Silent Mutation CTC,CTT L181L XP_005245802.1
XM_005245746.3 1032 Silent Mutation CTC,CTT L181L XP_005245803.1
XM_011540861.1 1032 Silent Mutation CTC,CTT L181L XP_011539163.1
XM_011540862.2 1032 Silent Mutation CTC,CTT L181L XP_011539164.1

View Full Product Details