Product Details

SNP ID
rs77990241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:11659911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTCAAGGCACAGACTGCACGG[A/G]ACCAGGTGGAATGCGGTCCAGGATT
Phenotype
MIM: 605538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NTSR2 PubMed Links

Gene Details

Gene
NTSR2
Gene Name
neurotensin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012344.3 Intron NP_036476.1
XM_005246156.3 Intron XP_005246213.1
XM_006711876.3 Intron XP_006711939.1
XM_006711877.3 Intron XP_006711940.1
XM_017003738.1 Intron XP_016859227.1

View Full Product Details