Product Details

SNP ID
rs76904124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:150470122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCCGCTTAACGTTTTTATTTGT[C/T]TTATTTACACATGCCAAGGTGGCAA
Phenotype
MIM: 602924
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RND3 PubMed Links

Gene Details

Gene
RND3
Gene Name
Rho family GTPase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001254738.1 878 Silent Mutation AAA,AAG K200K NP_001241667.1
NM_005168.4 878 Silent Mutation AAA,AAG K200K NP_005159.1

View Full Product Details