Product Details

SNP ID
rs76357989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:177219406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTTTATCCTGTATTATTCAAC[A/G]GGCTACAGTTCTTAGCAGGAGAGAG
Phenotype
MIM: 605372
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HNRNPA3 PubMed Links

Gene Details

Gene
HNRNPA3
Gene Name
heterogeneous nuclear ribonucleoprotein A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005246380.1 Intron XP_005246437.1
XM_005246381.4 Intron XP_005246438.1
XM_006712365.2 Intron XP_006712428.1
XM_017003587.1 Intron XP_016859076.1
XM_017003588.1 Intron XP_016859077.1
Gene
LOC105373759
Gene Name
uncharacterized LOC105373759
There are no transcripts associated with this gene.

Gene
MIR4444-1
Gene Name
microRNA 4444-1
There are no transcripts associated with this gene.

View Full Product Details