Product Details

SNP ID
rs76375624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206279461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAAACATGTCACCGTTCACAAT[A/G]TATCTGGATGTATTTATTTGGTCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZDBF2 PubMed Links

Gene Details

Gene
ZDBF2
Gene Name
zinc finger DBF-type containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285549.1 Intron NP_001272478.1
NM_020923.2 Intron NP_065974.1
XM_005246711.3 Intron XP_005246768.1
XM_005246712.4 Intron XP_005246769.1
XM_005246713.3 Intron XP_005246770.1
XM_006712656.3 Intron XP_006712719.1
XM_011511532.2 Intron XP_011509834.1
XM_011511533.2 Intron XP_011509835.1
XM_011511534.2 Intron XP_011509836.1
XM_011511535.2 Intron XP_011509837.1
XM_017004573.1 Intron XP_016860062.1
XM_017004574.1 Intron XP_016860063.1
XM_017004575.1 Intron XP_016860064.1
XM_017004576.1 Intron XP_016860065.1

View Full Product Details