Product Details

SNP ID
rs78030705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:222303976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCCACCGTGGGATCACGGGAACC[A/G]GAGGCAAGAGGCTGCGAATGAGTCC
Phenotype
MIM: 606597
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC140 PubMed Links

Gene Details

Gene
CCDC140
Gene Name
coiled-coil domain containing 140
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153038.1 458 Missense Mutation CAG,CGG Q25R NP_694583.1
XM_017003466.1 458 Missense Mutation CAG,CGG Q25R XP_016858955.1
Gene
PAX3
Gene Name
paired box 3
There are no transcripts associated with this gene.

View Full Product Details