Product Details

SNP ID
rs77314037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:224765791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGCTTTGCTAATTACTCGAGTG[C/T]AGGTTTGGTCCACTCCGCGCTTTGA
Phenotype
MIM: 611518
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOCK10 PubMed Links

Gene Details

Gene
DOCK10
Gene Name
dedicator of cytokinesis 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290263.1 6830 Missense Mutation TAC,TGC Y2158C NP_001277192.1
NM_014689.2 6830 Missense Mutation TAC,TGC Y2164C NP_055504.2
XM_005246673.4 6830 Missense Mutation TAC,TGC Y2177C XP_005246730.1
XM_006712617.3 6830 Silent Mutation CTA,CTG L2202L XP_006712680.1
XM_017004441.1 6830 Intron XP_016859930.1
XM_017004442.1 6830 Intron XP_016859931.1
XM_017004443.1 6830 Intron XP_016859932.1
XM_017004444.1 6830 Intron XP_016859933.1
XM_017004445.1 6830 Intron XP_016859934.1
XM_017004446.1 6830 Intron XP_016859935.1
XM_017004447.1 6830 Silent Mutation CTA,CTG L2193L XP_016859936.1

View Full Product Details