Product Details

SNP ID
rs80305648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227366120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTTTTTCAATGAAAATTCACAA[C/T]TGGCATTACTGTTGCTTGGAGAATT
Phenotype
MIM: 614785 MIM: 615404
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFF PubMed Links

Gene Details

Gene
MFF
Gene Name
mitochondrial fission factor
There are no transcripts associated with this gene.

Gene
TM4SF20
Gene Name
transmembrane 4 L six family member 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011511876.2 318 Missense Mutation AAT,AGT N58S XP_011510178.1

View Full Product Details