Product Details

SNP ID
rs380327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:65502255 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAGCAAAGTTTTGGGAAACTTTT[A/G]TCTTCCATTGGTGTTTTCAAGGTTA
Phenotype
MIM: 611502
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CENPK PubMed Links

Gene Details

Gene
CENPK
Gene Name
centromere protein K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267038.1 Intron NP_001253967.1
NM_022145.4 Intron NP_071428.2
XM_011543534.2 Intron XP_011541836.1
XM_011543535.2 Intron XP_011541837.1
XM_011543536.2 Intron XP_011541838.1
XM_011543537.2 Intron XP_011541839.1
XM_011543538.2 Intron XP_011541840.1
XM_011543539.2 Intron XP_011541841.1
XM_011543540.2 Intron XP_011541842.1
XM_011543541.2 Intron XP_011541843.1
XM_011543542.2 Intron XP_011541844.1
XM_017009690.1 Intron XP_016865179.1
XM_017009691.1 Intron XP_016865180.1
XM_017009692.1 Intron XP_016865181.1
XM_017009693.1 Intron XP_016865182.1
XM_017009694.1 Intron XP_016865183.1
XM_017009695.1 Intron XP_016865184.1
XM_017009696.1 Intron XP_016865185.1
XM_017009697.1 Intron XP_016865186.1
XM_017009698.1 Intron XP_016865187.1
XM_017009699.1 Intron XP_016865188.1
XM_017009700.1 Intron XP_016865189.1
XM_017009701.1 Intron XP_016865190.1

View Full Product Details