Product Details

SNP ID
rs724834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57786633 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAGAGAGTAGGTATTTTAGAGGA[C/G]GTTTGGGGGCCATAATTGGTGAAGA
Phenotype
MIM: 615258 MIM: 604723
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
METTL21B PubMed Links

Gene Details

Gene
METTL21B
Gene Name
methyltransferase like 21B
There are no transcripts associated with this gene.

Gene
TSFM
Gene Name
Ts translation elongation factor, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172695.1 Intron NP_001166166.1
NM_001172696.1 Intron NP_001166167.1
NM_001172697.1 Intron NP_001166168.1
NM_005726.5 Intron NP_005717.3

View Full Product Details