Product Details
- SNP ID
-
rs185647
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:130848145 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGGGAGCAGGCCATCTGCACAGG[G/A]TGGGTGACTTTGGCCTGAAGGGTGA
- Phenotype
-
MIM: 605216
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs55683027] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGEF4
- Gene Name
- Rho guanine nucleotide exchange factor 4
- Gene
- POTEI
- Gene Name
- POTE ankyrin domain family member I
View Full Product Details