Product Details

SNP ID
rs638562
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:59993396 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCACTCAATAATGTGAAATTTTAC[C/T]ATAGGATAATTATAATAATTATGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf87 PubMed Links

Gene Details

Gene
C1orf87
Gene Name
chromosome 1 open reading frame 87
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152377.2 Intron NP_689590.1
XM_017000307.1 Intron XP_016855796.1
XM_017000308.1 Intron XP_016855797.1

View Full Product Details