Product Details
- SNP ID
-
rs358327
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:140390008 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AAAACTGGTTTTTGGTTTATGCTGG[A/G]TTAAAAACAAAAAAGAAAAAAGACA
- Phenotype
-
MIM: 601858
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLGN
PubMed Links
Gene Details
- Gene
- CLGN
- Gene Name
- calmegin
- Gene
- SCOC
- Gene Name
- short coiled-coil protein
There are no transcripts associated with this gene.
View Full Product Details