Product Details

SNP ID
rs4613
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:240713870 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACACTCGGGTGTAAGACATCGGGAC[A/G]GGCGTCGTGACGTCGGGACGGCAGC
Phenotype
MIM: 601255
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF1A PubMed Links

Gene Details

Gene
KIF1A
Gene Name
kinesin family member 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244008.1 8728 UTR 3 NP_001230937.1
NM_001320705.1 8728 UTR 3 NP_001307634.1
XM_005247022.1 8728 UTR 3 XP_005247079.1
XM_005247024.1 8728 UTR 3 XP_005247081.1
XM_005247027.1 8728 UTR 3 XP_005247084.1
XM_005247028.2 8728 UTR 3 XP_005247085.1
XM_006712605.1 8728 UTR 3 XP_006712668.1
XM_011511364.1 8728 UTR 3 XP_011509666.1
XM_011511365.1 8728 UTR 3 XP_011509667.1
XM_011511366.2 8728 UTR 3 XP_011509668.1
XM_011511367.1 8728 UTR 3 XP_011509669.1
XM_017004383.1 8728 UTR 3 XP_016859872.1
XM_017004384.1 8728 UTR 3 XP_016859873.1
XM_017004385.1 8728 UTR 3 XP_016859874.1
XM_017004386.1 8728 UTR 3 XP_016859875.1
XM_017004387.1 8728 UTR 3 XP_016859876.1
XM_017004388.1 8728 UTR 3 XP_016859877.1
XM_017004389.1 8728 UTR 3 XP_016859878.1
XM_017004390.1 8728 UTR 3 XP_016859879.1
XM_017004391.1 8728 UTR 3 XP_016859880.1

View Full Product Details