Product Details

SNP ID
rs3886151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:119287839 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCTTACTTAAATTCCAATTTCTT[A/T]TAATATTAGAGTTGGTCTTCAGTTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C2orf76 PubMed Links

Gene Details

Gene
C2orf76
Gene Name
chromosome 2 open reading frame 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017927.3 Intron NP_001017927.2
NM_001322329.1 Intron NP_001309258.1
NM_001322330.1 Intron NP_001309259.1
NM_001322331.1 Intron NP_001309260.1
NM_001322332.1 Intron NP_001309261.1
XM_017003354.1 Intron XP_016858843.1

View Full Product Details