Product Details

SNP ID
rs235377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44888297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAGGATCTCCCAATCCCCTCAA[A/G]GGAGCCTCACGCAGCAACCTGACCG
Phenotype
MIM: 600065
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGB2 PubMed Links

Gene Details

Gene
ITGB2
Gene Name
integrin subunit beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000211.4 Intron NP_000202.3
NM_001127491.2 Intron NP_001120963.2
NM_001303238.1 Intron NP_001290167.1
XM_006724001.1 Intron XP_006724064.1
XM_017028341.1 Intron XP_016883830.1

View Full Product Details