Product Details
- SNP ID
-
rs34492112
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:75002576 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TCGATGCTGTTAGTGTCATGAACGC[G/A]TTAGTCCCGGTAGTGCCATCCTTTC
- Phenotype
-
MIM: 613766
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SCAMP5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs60407706] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SCAMP5
- Gene Name
- secretory carrier membrane protein 5
View Full Product Details