Product Details

SNP ID
rs12461478
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:51927486 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTGGGGGAGGGCCCAGAAGTGGAA[G/T]AATTCAGGAAAGTGCAGGTTCTGGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF613 PubMed Links

Gene Details

Gene
ZNF613
Gene Name
zinc finger protein 613
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031721.3 340 UTR 5 NP_001026891.2
NM_024840.3 340 UTR 5 NP_079116.2
XM_005259269.3 340 UTR 5 XP_005259326.1
XM_011527333.2 340 UTR 5 XP_011525635.1
XM_017027315.1 340 UTR 5 XP_016882804.1

View Full Product Details