Product Details

SNP ID
rs9674642
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76268223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTTTTCTTGAGAACTGGCCGAG[A/G]GGGCATATCTGGTTCAAAACCCTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
QRICH2 PubMed Links

Gene Details

Gene
QRICH2
Gene Name
glutamine rich 2
There are no transcripts associated with this gene.

Gene
UBALD2
Gene Name
UBA like domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182565.3 Intron NP_872371.1

View Full Product Details