Product Details

SNP ID
rs4778691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:82839526 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTAAAAACAGACACAAATATTTT[T/A]CTGAACTATGTATTTCTACAGAAAC
Phenotype
MIM: 604799 MIM: 612393
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
HOMER2 PubMed Links

Gene Details

Gene
HOMER2
Gene Name
homer scaffolding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004839.3 Intron NP_004830.2
NM_199330.2 Intron NP_955362.1
XM_005272448.4 Intron XP_005272505.1
XM_005272449.3 Intron XP_005272506.1
XM_006720775.3 Intron XP_006720838.1
XM_006720776.3 Intron XP_006720839.1
XM_011522231.2 Intron XP_011520533.1
XM_011522232.2 Intron XP_011520534.1
XM_011522233.2 Intron XP_011520535.1
XM_011522234.2 Intron XP_011520536.1
Gene
WHAMM
Gene Name
WAS protein homolog associated with actin, golgi membranes and microtubules
There are no transcripts associated with this gene.

View Full Product Details