Product Details

SNP ID
rs10061305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:170864857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGAGAAAAGGTATGTGGGAATG[A/G]CCATTCTGGCAGTGTGGACTTGAAT
Phenotype
MIM: 606141
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RANBP17 PubMed Links

Gene Details

Gene
RANBP17
Gene Name
RAN binding protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022897.4 Intron NP_075048.1
XM_011534627.1 Intron XP_011532929.1
XM_011534631.2 Intron XP_011532933.1
XM_011534636.2 Intron XP_011532938.1
XM_011534637.2 Intron XP_011532939.1
XM_017009736.1 Intron XP_016865225.1
XM_017009737.1 Intron XP_016865226.1
XM_017009738.1 Intron XP_016865227.1
XM_017009739.1 Intron XP_016865228.1
XM_017009740.1 Intron XP_016865229.1
XM_017009741.1 Intron XP_016865230.1
XM_017009742.1 Intron XP_016865231.1
XM_017009743.1 Intron XP_016865232.1
XM_017009744.1 Intron XP_016865233.1
XM_017009745.1 Intron XP_016865234.1
XM_017009746.1 Intron XP_016865235.1
XM_017009747.1 Intron XP_016865236.1

View Full Product Details