Product Details

SNP ID
rs11727729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55600709 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATAGAGGGAATCAGGGACTTGAA[C/T]AGAGAATTTTTGTATCATACCTGAA
Phenotype
MIM: 605103 MIM: 611676
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NMU PubMed Links

Gene Details

Gene
NMU
Gene Name
neuromedin U
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292045.1 Intron NP_001278974.1
NM_001292046.1 Intron NP_001278975.1
NM_006681.3 Intron NP_006672.1
XM_011534367.2 Intron XP_011532669.1
XM_011534368.2 Intron XP_011532670.1
Gene
PDCL2
Gene Name
phosducin like 2
There are no transcripts associated with this gene.

View Full Product Details