Product Details

SNP ID
rs11678309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10736296 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTATGACATGTTTGGACCTGGTAC[A/G]TTTTTAGTAGTTCTTTAATAATATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1C2 PubMed Links

Gene Details

Gene
ATP6V1C2
Gene Name
ATPase H+ transporting V1 subunit C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039362.1 Intron NP_001034451.1
NM_144583.3 Intron NP_653184.2
XM_011510339.2 Intron XP_011508641.1
XM_011510340.2 Intron XP_011508642.1
XM_011510341.2 Intron XP_011508643.1
XM_017003745.1 Intron XP_016859234.1

View Full Product Details