Product Details
- SNP ID
-
rs3780221
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:19059397 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTACTTAACTCTGCCATGATAGCA[C/T]GAAAGTAGCCATAAACAATGTCAAC
- Phenotype
-
MIM: 613433
MIM: 612194
MIM: 615646
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HAUS6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79496568] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HAUS6
- Gene Name
- HAUS augmin like complex subunit 6
- Gene
- RRAGA
- Gene Name
- Ras related GTP binding A
There are no transcripts associated with this gene.
- Gene
- SCARNA8
- Gene Name
- small Cajal body-specific RNA 8
There are no transcripts associated with this gene.
View Full Product Details