Product Details

SNP ID
rs13143747
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:44174943 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CACAGACTTTCTTTTTGGACAGATT[C/T]GTTTCCCGGGACTTGCTGATGAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD8 PubMed Links

Gene Details

Gene
KCTD8
Gene Name
potassium channel tetramerization domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198353.2 1553 Silent Mutation ACA,ACG T423T NP_938167.1
XM_011513690.2 1553 Silent Mutation ACA,ACG T451T XP_011511992.1

View Full Product Details