Product Details

SNP ID
rs11938536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70398474 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTAAGGGGGTAGACACACGCAC[A/C]TACAACTATAGGATCTGTCAACCTG
Phenotype
MIM: 608936 MIM: 611593
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OPRPN PubMed Links

Gene Details

Gene
OPRPN
Gene Name
opiorphin prepropeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302807.1 Intron NP_001289736.1
NM_021225.4 Intron NP_067048.4
Gene
SMR3B
Gene Name
submaxillary gland androgen regulated protein 3B
There are no transcripts associated with this gene.

View Full Product Details