Product Details

SNP ID
rs701820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97226715 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAACTAGTTACAGAGATAAGACAA[C/T]ATGTGAAGTAAATACAGTAAATCAT
Phenotype
MIM: 611587
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP19 PubMed Links

Gene Details

Gene
ARHGAP19
Gene Name
Rho GTPase activating protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204300.1 Intron NP_001191229.1
NM_001256423.1 Intron NP_001243352.1
NM_032900.5 Intron NP_116289.4
Gene
ARHGAP19-SLIT1
Gene Name
ARHGAP19-SLIT1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details