Product Details

SNP ID
rs6090295
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:63200865 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTTCATTAGCAATTATCCCTGCAA[A/C]TTTAGCAATTCAGTATTCATTTTAT
Phenotype
MIM: 616529
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
YTHDF1 PubMed Links

Gene Details

Gene
YTHDF1
Gene Name
YTH N6-methyladenosine RNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017798.3 Intron NP_060268.2

View Full Product Details