Product Details
- SNP ID
-
rs7966686
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52274932 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTGCAATGGGCTGGTGTGAACAG[G/T]TCTTGCTCTCTGGGTTGAGGGGGTG
- Phenotype
-
MIM: 601928
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT86
PubMed Links
Gene Details
- Gene
- KRT86
- Gene Name
- keratin 86
View Full Product Details