Product Details
- SNP ID
-
rs13231520
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:5601915 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGAAGGGGACGTGTAGGACTTCATA[A/G]TTTCCTTTTTTTTGTTTTAGAGACA
- Phenotype
-
MIM: 602689
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FSCN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200277948] are located under a probe and SNP(s) [rs57071890] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FSCN1
- Gene Name
- fascin actin-bundling protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003088.3 |
|
Intron |
|
|
NP_003079.1 |
- Gene
- LOC107986762
- Gene Name
- uncharacterized LOC107986762
There are no transcripts associated with this gene.
View Full Product Details