Product Details

SNP ID
rs7451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:64632545 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAACAAGGGTGCCCATGTGGTTGC[C/T]CCTTCTGTCATCCAGAGAAAGTGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERTAD2 PubMed Links

Gene Details

Gene
SERTAD2
Gene Name
SERTA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014755.2 4678 UTR 3 NP_055570.1
XM_005264669.2 4678 UTR 3 XP_005264726.1
XM_011533204.1 4678 UTR 3 XP_011531506.1
XM_011533205.2 4678 UTR 3 XP_011531507.1

View Full Product Details