Product Details
- SNP ID
-
rs11807921
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.1:220754675 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACTAAGCAGTGAGGGAGAGTCTCT[C/G]AGCCAGGCTCATGTTCTTGTTTTTG
- Phenotype
-
MIM: 614127
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MARC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201384609] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MARC2
- Gene Name
- mitochondrial amidoxime reducing component 2
View Full Product Details