Product Details

SNP ID
rs1390082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73516595 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAAGAACTCCATCTTAAAAAAAA[A/T]TTTTAAAAACCATCACACAAACAGA
Phenotype
MIM: 130590
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EEF1A1 PubMed Links

Gene Details

Gene
EEF1A1
Gene Name
eukaryotic translation elongation factor 1 alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001402.5 2667 UTR 3 NP_001393.1
XM_011535514.2 2667 Intron XP_011533816.1

View Full Product Details