Product Details

SNP ID
rs2270576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48930601 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCCAGCCACGCCAACCCTTCCTT[C/T]AGCAGGTGTTCCTGGAGGGAAAAGG
Phenotype
MIM: 610904 MIM: 611362
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNF8 PubMed Links

Gene Details

Gene
SNF8
Gene Name
SNF8, ESCRT-II complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317192.1 756 Silent Mutation CTA,CTG L216L NP_001304121.1
NM_001317193.1 756 Silent Mutation CTA,CTG L200L NP_001304122.1
NM_001317194.1 756 Silent Mutation CTA,CTG L113L NP_001304123.1
NM_007241.3 756 Silent Mutation CTA,CTG L217L NP_009172.2
XM_017024075.1 756 Silent Mutation CTA,CTG L113L XP_016879564.1
Gene
UBE2Z
Gene Name
ubiquitin conjugating enzyme E2 Z
There are no transcripts associated with this gene.

View Full Product Details