Product Details

SNP ID
rs2844337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77322663 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGCATGGGAACACTATTGGGTCA[A/C]GGTCCTCCCGAACAGCTGCTAGAAG
Phenotype
MIM: 602590
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PAK1 PubMed Links

Gene Details

Gene
PAK1
Gene Name
p21 (RAC1) activated kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128620.1 2498 UTR 3 NP_001122092.1
NM_002576.4 2498 UTR 3 NP_002567.3
XM_011545080.2 2498 UTR 3 XP_011543382.1
XM_011545081.2 2498 Intron XP_011543383.1
XM_011545082.2 2498 Intron XP_011543384.1
XM_011545083.2 2498 Intron XP_011543385.1
XM_011545084.2 2498 Intron XP_011543386.1
XM_017017846.1 2498 UTR 3 XP_016873335.1
XM_017017847.1 2498 UTR 3 XP_016873336.1
XM_017017848.1 2498 UTR 3 XP_016873337.1
XM_017017849.1 2498 UTR 3 XP_016873338.1
XM_017017850.1 2498 UTR 3 XP_016873339.1

View Full Product Details