Product Details

SNP ID
rs3790046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94052640 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGGTAGTTGACTACTTCCATTTT[A/G]TGGAGGAGAAAGCTGAGAACACAAC
Phenotype
MIM: 606181 MIM: 608338
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX24 PubMed Links

Gene Details

Gene
DDX24
Gene Name
DEAD-box helicase 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020414.3 Intron NP_065147.1
Gene
OTUB2
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 2
There are no transcripts associated with this gene.

View Full Product Details