Product Details

SNP ID
rs2029984
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52007011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTGCCCTTTGTAAGCTATGTCT[A/G]TAATTACTGAGGCAACCTTTTAACC
Phenotype
MIM: 610858 MIM: 605399
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C14orf166 PubMed Links

Gene Details

Gene
C14orf166
Gene Name
chromosome 14 open reading frame 166
There are no transcripts associated with this gene.

Gene
NID2
Gene Name
nidogen 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007361.3 Intron NP_031387.3
XM_005267405.4 Intron XP_005267462.1
XM_005267406.4 Intron XP_005267463.1
XM_005267407.4 Intron XP_005267464.1

View Full Product Details